Learn More
Memmert™ Potential-Free Contact
For combination error message (e.g. supply failure,sensor fault, fuse)
Brand: Memmert™ IPP1400ecoH6
3357.55 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- For combination error message (e.g. supply failure,sensor fault, fuse)
Specifications
Combination error message | |
IPP1400eco |
Product Content Correction
Product Title
Memmert™ Potential-Free Contact > IPP1400eco