missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tin(IV) oxide trihydrate, 95%, Thermo Scientific Chemicals
561.60 NOK - 1566.00 NOK
Chemical Identifiers
CAS | 12125-03-0 |
---|---|
Molecular Formula | H6K2O6Sn |
Molecular Weight (g/mol) | 298.95 |
MDL Number | MFCD00150391 |
InChI Key | HTHDWDSBYOUAFF-UHFFFAOYSA-N |
Synonym | potassium stannate trihydrate, unii-r38y86o4sy, 2k.so3n.3h2o, potassium tin iv oxide trihydrate, dipotassium dioxido oxo tin trihydrate, dipotassium ion oxostannanebis olate trihydrate, dipotassium oxostannanebis olate trihydrate, dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate |
PubChem CID | 18694711 |
IUPAC Name | dipotassium;dioxido(oxo)tin;trihydrate |
SMILES | O.O.O.[K+].[K+].[O-][Sn]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311549
|
Thermo Scientific Alfa Aesar
014015.30 |
250 g |
561.60 NOK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321549
|
Thermo Scientific Alfa Aesar
014015.A1 |
1 kg |
1566.00 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium tin(IV) oxide is a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12125-03-0 | |
298.95 | |
HTHDWDSBYOUAFF-UHFFFAOYSA-N | |
18694711 | |
O.O.O.[K+].[K+].[O-][Sn]([O-])=O |
H6K2O6Sn | |
MFCD00150391 | |
potassium stannate trihydrate, unii-r38y86o4sy, 2k.so3n.3h2o, potassium tin iv oxide trihydrate, dipotassium dioxido oxo tin trihydrate, dipotassium ion oxostannanebis olate trihydrate, dipotassium oxostannanebis olate trihydrate, dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate | |
dipotassium;dioxido(oxo)tin;trihydrate |
Specifications
140°C (decomposition) | |
Crystalline | |
MFCD00150391 | |
Soluble in water. | |
O.O.O.[K+].[K+].[O-][Sn]([O-])=O | |
298.95 | |
298.94 (244.89 Anhydrous) | |
Odorless | |
Potassium tin(IV) oxide trihydrate |
12125-03-0 | |
H6K2O6Sn | |
potassium stannate trihydrate, unii-r38y86o4sy, 2k.so3n.3h2o, potassium tin iv oxide trihydrate, dipotassium dioxido oxo tin trihydrate, dipotassium ion oxostannanebis olate trihydrate, dipotassium oxostannanebis olate trihydrate, dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate | |
HTHDWDSBYOUAFF-UHFFFAOYSA-N | |
dipotassium;dioxido(oxo)tin;trihydrate | |
18694711 | |
≥95% | |
(metals basis) |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only