missing translation for 'onlineSavingsMsg'
Learn More
Potassium Phosphate Monobasic, MP Biomedicals™ Product Code.: 11440562

Potassium Phosphate Monobasic, MP Biomedicals™
BIOPHARMA

Product Code. 11440562 Shop All MP Biomedicals Products
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11440562

Brand: MP Biomedicals 02191430.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a component for a wide variety of media used in the culture of microorganisms. Potassium Phosphate Monobasic, MP Biomedicals is a reagent with high buffering capacity that helps maintain pH and supplies essential phosphate. It is a component in ph

Potassium phosphate occurs in several forms: monobasic, dibasic, and tribasic (K3PO4). Most pH-neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7778-77-0
Physical Form Powder
Quantity 500 g
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
Solubility Information Soluble in water (0.5G in 10mL water), Insoluble to slightly soluble in ethanol; insoluble in alcohol.
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.09
Grade ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS6836 Certificate TypeCertificate of Analysis Date17/03/2022 Catalog Number02191430.5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Phosphate Monobasic, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.