missing translation for 'onlineSavingsMsg'
Learn More
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals Product Code.: 11430562

Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
BIOPHARMA

Product Code. 11430562 Shop All MP Biomedicals Products
100 g
Click to view available options
Quantity:
100 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11430562

Brand: MP Biomedicals 02191430.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow MoreShow Less
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Quantity 100 g

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS6836 Certificate TypeCertificate of Analysis Date17/03/2022 Catalog Number02191430.1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.