Learn More
Potassium phosphate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1340°C |
CAS | 7778-53-2 |
Physical Form | Solid |
Quantity | 100 g |
Molecular Formula | K{3}O{4}P |
Synonym | Potassium phosphate, tribasic;Tripotassium phosphate |
Percent Purity | 97% |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0468878 | Certificate of Analysis | 17/05/2025 | L15168.36,L15168.22,L15168.0P |
A0460235 | Certificate of Analysis | 11/11/2024 | L15168.36,L15168.0P,L15168.22 |
10252025 | Certificate of Analysis | 14/08/2024 | L15168.36,L15168.0P,L15168.22 |
A0438936 | Certificate of Analysis | 21/02/2024 | L15168.36,L15168.0P,L15168.22 |
10238939 | Certificate of Analysis | 24/02/2023 | L15168.36,L15168.0P,L15168.22 |
Safety and Handling


- Potassium phosphate
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Phosphoric acid, tripotassium salt
Your input is important to us. Please complete this form to provide feedback related to the content on this product.