missing translation for 'onlineSavingsMsg'
Learn More

Potassium phenoxymethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Product Code. 15404707
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027642-30-3
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD10566516
InChI Key OVYSNXCRBZPJIG-UHFFFAOYSA-N
Synonym potassium phenoxy-methyltrifluoroborate, potassium trifluoro phenoxymethyl boranuide, pubchem11600, potassium phenoxymethyltrifluoroborate, potassium trifluoro phenoxymethyl borate, potassium trifluoro phenoxymethyl borate 1-, potassium phenoxymethyltrifluoroborate, contains kbr
PubChem CID 45479874
IUPAC Name potassium;trifluoro(phenoxymethyl)boranuide
SMILES [B-](COC1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Quantity 250 mg
Formula Weight 214.04
Percent Purity 95%
Chemical Name or Material Potassium phenoxymethyltrifluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'