Learn More
Potassium perrhenate, 99% (metals basis), Re 64%, Thermo Scientific Chemicals
Description
A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1360°C to 1370°C |
Melting Point | 550°C |
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN1479 |
Solubility Information | Soluble in water. |
Formula Weight | 289.3 |
Percent Purity | 99% |
Concentration or Composition (by Analyte or Components) | Re 64% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M20L004 | Certificate of Analysis | 22/01/2025 | 011411.18,011411.09,011411.04 |
10184220 | Certificate of Analysis | 17/10/2022 | 011070.A1,011411.18,011070.30,011070.18,011070.D9 |
10199907 | Certificate of Analysis | 17/10/2022 | 011070.A1,011411.18,011070.30,011070.18,011070.D9 |
Z07H023 | Certificate of Analysis | 12/04/2022 | 011411.18,011411.09,011411.04,011421.14,011411.SS |
Z17H025 | Certificate of Analysis | 12/04/2022 | 011070.A1,011411.18,011070.30,011070.18,011070.D9 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.