missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, (Crystalline/ACS Reagent Grade), MP Biomedicals
2285.00 NOK - 3495.00 NOK
Chemical Identifiers
CAS | 7757-79-1 |
---|---|
Molecular Formula | KNO3 |
Molecular Weight (g/mol) | 101.10 |
MDL Number | MFCD00011409 |
InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitrasShow More |
PubChem CID | 24434 |
ChEBI | CHEBI:63043 |
IUPAC Name | potassium nitrate |
SMILES | [K+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480552
|
MP Biomedicals
0219142801 |
1 kg |
2285.00 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490552
|
MP Biomedicals
0219142805 |
5 kg |
3495.00 NOK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7757-79-1 | |
101.10 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
24434 | |
potassium nitrate |
KNO3 | |
MFCD00011409 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitrasShow More | |
CHEBI:63043 | |
[K+].[O-][N+]([O-])=O |
Specifications
7757-79-1 | |
KNO3 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
CHEBI:63043 | |
ACS Reagent |
Crystalline | |
MFCD00011409 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium nitrate | |
24434 | |
101.1 | |
Potassium Nitrate |
Safety and Handling
missing translation for 'storageNote' : Not for export
Recommended Storage : Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Nitrate, (Crystalline/ACS Reagent Grade), MP Biomedicals