missing translation for 'onlineSavingsMsg'
Learn More
Potassium Nitrate, ACS Reagent Grade, Scharlab Product Code.: 11304274
compliance-icons

Potassium Nitrate, ACS Reagent Grade, Scharlab

Product Code. 11304274
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11304274

Brand: Scharlab PO02870500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, an oxidizing agent, in pyrotechnics, in the manufacture of glass and in food industry.

  • Synonyms: Nitric acid potassium salt, Saltpeter
  • Molecular Formula: KNO3
  • Molar Mass: 101.11g/mol
  • CAS: 7757-79-1
  • EIN ECS No.: 231-818-8
  • Iodates (IO3): 0.0005% max
  • Nitrites (NO2): 0.001%

Specifications

Melting Point 334°C
CAS 7757-79-1
pH 4.5–8.5 (5%, water)
Quantity 500 g
Molecular Formula KNO3
MDL Number MFCD00011409
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium;nitrate
Molecular Weight (g/mol) 101.10
PubChem CID 24434
ChEBI CHEBI:63043
Formula Weight 101.11g/mol
Percent Purity 99% min (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Potassium nitrate, reagent grade, ACS, ISO Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Nitrate, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.