Learn More
Potassium nitrate, 99%, extra pure Product Code.: 10189909
Potassium nitrate, 99%, extra pure
Product Code. 10189909 Shop All Thermo Scientific Chemicals Products
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Product Code. 10189909
Brand: Thermo Scientific Acros 434430010
Request Bulk or Custom Format
CAS: 7757-79-1 | KNO3 | 101.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitrasShow MoreShow Less
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O
Specifications
Boiling Point 400.0°C
Melting Point 334.0°C
Color White
Physical Form Fine Crystals or Crystalline Powder
Quantity 1 kg
Assay Percent Range 99%
Linear Formula KNO3
Merck Index 15,7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
IUPAC Name potassium;nitrate
Formula Weight 101.1
Percent Purity 99%
Grade Extra Pure
Packaging Plastic Bottle
Chemical Name or Material Potassium nitrate, Extra Pure
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453213 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number434430025,434430100,434430010
Lot NumberA0410381 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number434430025,434430100,434430010
Lot NumberA0408688 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number434430025,434430100,434430010
Lot NumberA0405913 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number434430025,434430100,434430010
Lot NumberA0408687 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number434430025,434430100,434430010
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use sand to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate
Product Title
Potassium nitrate, 99%, extra pure >