missing translation for 'onlineSavingsMsg'
Learn More
Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11308308

Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11308308
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11308308

Brand: Thermo Scientific Alfa Aesar 039501.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Employed as corrosion inhibitors and anti-scaling agents. Used widely in chemical fertilizer industries, glass and ceramic industries, pharmaceutical industries. Also used in the process of pigment production for plastics, enamels, ceramics. Used for industrial gas desulphuration and H2S adsorption and elimination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13769-43-2
Molecular Formula KO3V
Molecular Weight (g/mol) 138.04
MDL Number MFCD00050038
InChI Key BQFYGYJPBUKISI-UHFFFAOYSA-N
Synonym potassium metavanadate, potassium vanadate, potassium m-vanadate, potassium trioxovanadate, potassium vanadium trioxide, unii-0s0516d6vi, vanadate vo31-, potassium, vanadic acid, potassium salt, potassium vanadate v kvo3 6ci,7ci, potassium dioxidovanadium 1+Show MoreShow Less
PubChem CID 4571336
IUPAC Name potassium;oxido(dioxo)vanadium
SMILES [K+].[O-][V](=O)=O

Specifications

Melting Point 520°C
Physical Form Powder
Quantity 100 g
UN Number UN2864
Solubility Information Soluble in water.
Formula Weight 138.04
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium metavanadate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX08K047 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number039501.14,039501.22
Lot NumberU06K002 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number039501.14,039501.22
Lot NumberM03K005 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number039501.14,039501.22
Lot NumberY01E018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039501.14,039501.22
Lot NumberD09M42 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039501.14,039501.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.