missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, ACS Reagent Grade, Scharlab

Product Code. 11334254
1 kg, 1kg
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent and as an emulsifier in photography.

  • Synonym: Knollide
  • Molecular Formula: KI
  • Molar Mass: 166.01g/mol
  • CAS: 7681-11-0
  • EIN ECS No.: 231-659-4
  • Reducing Substances: Passes test

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 686°C
pH 6.0–9.2 (5%, water)
Quantity 1 kg
Solubility Information Soluble in water (20°C)
Formula Weight 166.01g/mol
Percent Purity 99.5% min (argentometric)
Grade ACS Reagent
Loss on Drying 0.2% max (150°C, 6h)
Vapor Pressure 1.3 hPa (at 745°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Iodide, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.