Learn More
Macherey-Nagel™ Potassium Iodate Starch Paper
Brand:  Macherey-Nagel™ 90753
Product Code. 11346544
  • 194.00 NOK / 5m
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
White to Blue/Black
Determination of Sulphit and Ssulphur dioxide
Qualitative Test Paper
7 mm
Reel
1 Each
5000 mm