missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 45%, for HPLC, Honeywell Fluka™

for HPLC, ∼45%

463.96 NOK - 1861.51 NOK

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15698290
SDS View Documents
Honeywell-Fluka
03564-25ML
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15670680
SDS View Documents
Honeywell-Fluka
03564-4X25ML
4 x 25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15660680
SDS View Documents Promotion Details
Honeywell-Fluka
03564-100ML
100 mL
478.01 NOK
100mL
Please to purchase this item. Need a web account? Register with us today!
15680680
SDS View Documents
Honeywell-Fluka
03564-500ML
500 mL
1861.51 NOK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310

H290-H302-H314

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Hydroxide Solution, 45%, for HPLC, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.