missing translation for 'onlineSavingsMsg'
Learn More

Potassium heptafluorotantalate(V), 99.99% (metals basis)

Product Code. p-7058996
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11370348

Brand: Thermo Scientific Alfa Aesar 043733.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16924-00-8 | F7K2Ta | 392.133 g/mol

It is an important raw material used as a primary and secondary pharmaceutical intermediate and also used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16924-00-8
Molecular Formula F7K2Ta
Molecular Weight (g/mol) 392.133
MDL Number MFCD00042369
InChI Key DPXAUKPQZOAHGF-UHFFFAOYSA-G
Synonym potassium heptafluorotantalate, potassium tantalum fluoride, dipotassium heptafluorotantalate, tantalum potassium fluoride, potassium fluotantalate, potassium heptafluorotantalate v, potassium fluorotantalate, dipotassium heptafluorotantalum 2-, potassium fluorotanatalate k2taf7, potassium tantalum fluoride k2taf5
PubChem CID 28146
IUPAC Name dipotassium;heptafluorotantalum(2-)
SMILES F[Ta-2](F)(F)(F)(F)(F)F.[K+].[K+]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information Decomposes in water. Slightly soluble in hydrogen fluoride.
Formula Weight 392.14
Percent Purity 99.99%
Density 4.560 g/mL
Assay (metals basis)
Chemical Name or Material Potassium heptafluorotantalate (V)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.