missing translation for 'onlineSavingsMsg'
Learn More
Potassium Dihydrogen Phosphate, ACS Reagent Grade, Scharlab Product Code.: 11334234

Potassium Dihydrogen Phosphate, ACS Reagent Grade, Scharlab

Product Code. 11334234
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11334234

Brand: Scharlab PO02601000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in analytical chemistry, in buffer solutions.

  • Synonyms: Potassium biphosphate, Potassium phosphate monobasic, Primary potassium phosphate, Mono-potassium phosphate
  • Molecular Formula: KH2PO4
  • Molar Mass: 136.09g/mol
  • CAS: 7778-77-0
  • EIN ECS No.: 231-913-4

Specifications

Melting Point ∼253°C (decomposition)
CAS 7778-77-0
pH 4.2–4.5 (5%, water)
Quantity 1 kg
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.09g/mol
Percent Purity 99.5% min (acidimetric)
Grade ACS Reagent
Identification Passes Test
Loss on Drying 0.2% max (105°C, 1h)
Appearance Clear and colorless
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Dihydrogen Phosphate, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.