Learn More
Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10532533
Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Product Code. 10532533 Shop All Fisher Chemical Products
1 AMP
Click to view available options
Quantity:
1 AMP
6 AMP
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10532533
Brand: Fisher Chemical J/6212C/05
Request Bulk or Custom Format
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Cr2K2O7, CAS Number-7778-50-9, dipotassium bichromate, bichromate of potash, dichromic acid dipotassium salt, potassium dichromate, potassium dichromate vi, iopezite, dipotassium dichromium heptaoxide, kaliumdichromat, potassium bichromate, dipotassium dichromate, 1AMP, CHEBI:53444, Orange
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
MDL Number 11367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potashShow MoreShow Less
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Specifications
Color Orange
Physical Form Liquid
Quantity 1 AMP
Packaging Ampoule
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium dichromate, solution 0.0167 M, (0.1N), concentrated solution
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Potassium dichromate
Product Content Correction
Product Title
Potassium Dichromate Solution 0.0167M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™ >