missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Cyanide, puriss. p.a. ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 250 g |
Linear Formula | KCN |
UN Number | 1680 |
Beilstein | 3593645 |
Formula Weight | 65.12g/mol |
Percent Purity | ≥97% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Potassium Cyanide, Reag. Ph. Eur. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction