missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate solution 0.033mol/L, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
Concentration | 0.0333M (0.1N) |
CAS | 7732-18-5 |
Packaging | Bottle |
Quantity | 1000 mL |
Chemical Name or Material | Potassium Chromate |
Safety and Handling
Product Identifier
- Potassium chromate 1/30 - 0.0333M (0.1N) Volumetric Solution
- Danger
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H317-May cause an allergic skin reaction.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction