Learn More
Potassium Chloride Solution 3M, Reagecon™
BIOPHARMA
Brand:  Reagecon EFS35
Quantity 500 mL
Product Code. 11553162
  • 733.00 NOK / 500mL
Specifications
3 mol/L
500 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberEFS324L1 Certificate TypeCertificate of Analysis Date31/05/2025 Catalog NumberEFS35
Lot NumberEFS324G1 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberEFS35
Lot NumberEFS324B1 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog NumberEFS35
Lot NumberEFS323L1 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog NumberEFS35
Lot NumberEFS323D1 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog NumberEFS35
Promotions
Promotions
Product Content Correction
Product Title
Potassium Chloride Solution 3M, Reagecon™ > Quantity: 500mL