missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride, MP Biomedicals™ Product Code.: 11420552

Potassium Chloride, MP Biomedicals™

Product Code. 11420552 Shop All MP Biomedicals Products
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11420552

Brand: MP Biomedicals 02191427.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the preparation of phosphate buffered saline (PBS) and for the extraction and solubilization of proteins. Potassium Chloride, MP Biomedicals is a good potassium source widely used as a reagent in biochemistry and molecular biology.

Widely used reagent in biochemistry and molecular biology:

  • In the preparation of phosphate buffered saline (PBS)
  • For the extraction and solubilization of proteins
  • In the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes
  • For dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M)
  • In the preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Soluble in water (1g/3mL, clear, colorless solution), boiling water (1g/1.8mL), glycerol (1g/14mL), or ethanol (1g/250mL); insoluble in ether or acetone; hydrochloric acid, sodium or magnesium chlorides diminish its solubility in water.
Formula Weight 74.55
Percent Purity 99.0 to 100.5%
Grade ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.