missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals Product Code.: 11450552

Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11450552 Shop All MP Biomedicals Products
5 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11450552

Brand: MP Biomedicals 0219142705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Physical Form Crystalline
Quantity 5 kg
Formula Weight 74.55
Grade ACS Reagent
Chemical Name or Material Potassium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.