missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, ReagentPlus™, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
12 kg
Unit Size:
12kg
1kg
2.50kg
5kg
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Quantity | 1 kg |
Linear Formula | K2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4267587 |
IUPAC Name | dipotassium carbonate |
Formula Weight | 138.21g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Chemical Name or Material | Potassium Carbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Carbonate, ReagentPlus™, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction