Learn More
Fisherbrand™ Porcelain Plate For Desiccators
For use with desiccators
268.00 NOK - 1515.00 NOK
Product Code | Brand | For Use With (Equipment) | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
15594645
|
Fisherbrand™
9180003209 |
Desiccator (Dia.: 250mm) | 235 mm |
1115.00 NOK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
15584645
|
Fisherbrand™
9180003208 |
Desiccator (Dia.: 200mm) | 190 mm |
805.00 NOK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
15574645
|
Fisherbrand™
9180003207 |
Desiccator (Dia.: 150mm) | 140 mm |
414.00 NOK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
15564645
|
Fisherbrand™
9180003206 |
Desiccator (Dia.: 100mm) | 90 mm |
268.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
15504655
|
Fisherbrand™
9180003210 |
Desiccator (Dia.: 300mm) | 280 mm |
1515.00 NOK
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
Description
- For use with desiccators
Specifications
Plate |
Product Title
Fisherbrand™ Porcelain Plate For Desiccators