Learn More
Hecht Karl™ Porcelain Pestle
157.00 NOK - 273.00 NOK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10489061
|
Hecht Karl™
47031013 |
130mm |
273.00 NOK
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
10309661
|
Hecht Karl™
47031006 |
60mm |
157.00 NOK
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
10064051
|
Hecht Karl™
47031008 |
80mm |
224.00 NOK
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
Specifications
Pestle | |
Porcelain | |
Pestle |