Learn More
Haldenwanger™ Porcelain Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
125.00 NOK - 285.00 NOK
Specifications
Material | Porcelain |
---|---|
Shape | Rectangular |
Type | Evaporating Basin |
For Use With (Application) | Laboratories |
Interior Finish | Smooth |
Product Code | Brand | Height (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
10509031
|
Haldenwanger™
L 33C/00 |
10 mm | 45 mm |
125.00 NOK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
10048160
|
Haldenwanger™
L 33C/4B |
13 mm | 70 mm |
211.00 NOK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
10260961
|
Haldenwanger™
L 33C/00B |
15 mm | 50 mm |
177.00 NOK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
10038150
|
Haldenwanger™
L 33C/7 |
16 mm | 96 mm |
285.00 NOK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Shallow form, round-bottom porcelain basin
- Available in a variety of sizes
- Includes spout for easy pouring
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
Evaporating Basin | |
Smooth |
Rectangular | |
Laboratories |
Product Title
Haldenwanger™ Porcelain Basins