Learn More
Applied Biosystems™ POP-6™ Polymer, for 3500/SeqStudio™ Flex Product Code.: 15879826
Applied Biosystems™ POP-6™ Polymer, for 3500/SeqStudio™ Flex
Product Code. 15879826 Shop All Applied Biosystems Products
384 Samples, Each
Click to view available options
Quantity:
384 Samples
96 Samples
960 Samples
Unit Size:
Each
Pack of 384
Pack of 96
Pack of 960
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 15879826
Brand: Applied Biosystems™ 4393717
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Includes
The pouch contains 384 samples of the polymer
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-6 separation matrix is optimized for standard sequencing and fragment analysis.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-6 separation matrix is optimized for standard sequencing and fragment analysis. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-6 separation matrix is optimized for standard sequencing and fragment analysis.
• Easy to use: Pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: alow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID/Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Quantity 384 Samples
For Use With (Application) Standard Sequencing, Fragment Analysis (Denaturing)
Product Line POP-6™
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2410188 Certificate TypeCertificate of Analysis Date21/01/2025 Product Code4393717
Lot Number2410187 Certificate TypeCertificate of Analysis Date03/12/2024 Product Code4393717
Lot Number2409186 Certificate TypeCertificate of Analysis Date01/11/2024 Product Code4393717
Lot Number2409185 Certificate TypeCertificate of Analysis Date03/10/2024 Product Code4393717
Lot Number2408184 Certificate TypeCertificate of Analysis Date16/09/2024 Product Code4393717
  • Multiple Manufacturers
    Exclusive savings, limited-time offer.
    Promo Code: 24112
    Expires2025-06-27
    New Offer
    Save Now
Product Content Correction
Product Title
Applied Biosystems™ POP-6™ Polymer, for 3500/SeqStudio™ Flex >