Learn More
Applied Biosystems™ POP-4™ Polymer, for 3500/SeqStudio™ Flex Product Code.: 15829876
Applied Biosystems™ POP-4™ Polymer, for 3500/SeqStudio™ Flex
Product Code. 15829876 Shop All Applied Biosystems Products
96 Samples, Each
Click to view available options
Quantity:
384 Samples
96 Samples
960 Samples
Unit Size:
Each
Pack of 384
Pack of 96
Pack of 960
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 15829876
Brand: Applied Biosystems™ A26070
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Includes
The pouch contains 384 samples of the polymer
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-4 separation matrix is optimized for HID/forensic applications.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-4 separation matrix is optimized for HID/forensic applications. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-4 separation matrix is optimized for HID/forensic applications. 
• Easy to use: Pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: alow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID/Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
96 Samples
TRUSTED_SUSTAINABILITY
Specifications
Quantity 96 Samples
For Use With (Application) Human Identification
Product Line POP-4™
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2410096 Certificate TypeCertificate of Analysis Date24/01/2025 Product CodeA26070
Lot Number2410095 Certificate TypeCertificate of Analysis Date29/10/2024 Product CodeA26070
Lot Number2409094 Certificate TypeCertificate of Analysis Date07/10/2024 Product CodeA26070
Lot Number2408093 Certificate TypeCertificate of Analysis Date29/08/2024 Product CodeA26070
Lot Number2406092 Certificate TypeCertificate of Analysis Date05/08/2024 Product CodeA26070
Product Content Correction
Product Title
Applied Biosystems™ POP-4™ Polymer, for 3500/SeqStudio™ Flex >