missing translation for 'onlineSavingsMsg'
Learn More
Polyvinyl Alcohol, White to off-white powder, MP Biomedicals Product Code.: 11467182

Polyvinyl Alcohol, White to off-white powder, MP Biomedicals

Product Code. 11467182 Shop All MP Biomedicals Products
250 g, 250g
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 11467182

Brand: MP Biomedicals 0215193883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 9002-89-5
Color White
Density 1.25-1.50g/mL
Assay Percent Range 4%
Molecular Formula (C2H4O)n
MDL Number MFCD00081922
Quantity 250 g
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES OC(-*)C-*
IUPAC Name ethenol
Molecular Weight (g/mol) 44.05
PubChem CID 11199
Formula Weight 22000
Physical Form Powder
Chemical Name or Material Polyvinyl Alcohol, Aqueous Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyvinyl Alcohol, White to off-white powder, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.