Learn More
Megro™ Polystyrene Tweezer
Single packed disposable sterile forceps are ideal for dissection use and delicate tasks.
Brand:  Megro™ 126300
Product Code. 10052199
  • 293.61 NOK / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Order Info
dimLengthMetric
Specifications
Specifications
50Pack
Polystyrene
Tweezers
Anatomical
125 mm
Each
Product Title
Megro™ Polystyrene Tweezer > Length: 125mm