Learn More
Brand™ Disposable Polystyrene Stirring Spatulas
Disposable stirring spatula made of polystyrene material with a L x diam. of 120 mm x 3 mm that can be used in clinical, scientific, research, and other types of laboratories.
Brand: Brand™ 759800
135.79 NOK valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
- Disposable stirring spatula
- Made of polystyrene material
- Can be used in clinical, scientific, research, and other types of laboratories
- L × diam. 120 mm × 3 mm
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
500 Pk. | |
Stirring Spatula | |
120 mm |
Single-use | |
Polystyrene |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24154Expires: 2025-02-28
Product Content Correction
Product Title
Brand™ Disposable Polystyrene Stirring Spatulas > Diameter: 3mm