Learn More
TPP™ Polystyrene Petri Dishes
Manufactured from crystal clear polystyrene to provide visual control of cells in stacked dishes using transmitted light. TPP™ Polystyrene Petri Dishes are designed with a gripping ring for easy handling and to prevent accidental dropping of dish bottom.
1425.00 NOK - 1740.00 NOK
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
Shape | Round |
Sterility | Sterile |
Type | Petri Dish |
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
11361774
|
TPP™
93100 |
96 mm | 17 mL | 60 sq. cm |
1740.00 NOK
Pack of 240 |
In Stock
Log in to see stock. ![]() |
||||
11371774
|
TPP™
93150 |
146 mm | 42 mL | 147 sq. cm |
1425.00 NOK
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Manufactured from crystal clear polystyrene to provide visual control of cells in stacked dishes using transmitted light. TPP™ Polystyrene Petri Dishes are designed with a gripping ring for easy handling and to prevent accidental dropping of dish bottom.
- Surface treatment of the growth area optimally enhances proliferation of cells
- Serrations in the gripping ring render the dish grip secure
- Yellow marking area on side of the lid, frosted on side of the bottom
- Stacking ring on the lid and corresponding ridge on the base provide secure stacking
- Consistent growth conditions:
- Vents provide airflow between stacked dishes for consistent growth and prevention of condensation
- Cams on the inside of the dish lid function as spacer, limit condensation and enable a constant movement of air
- Numeric "clock" markings (12, 3, 6, 9) in the base of the dish divide it into quadrants for quick orientation
Specifications
Clear | |
Round | |
Petri Dish |
Polystyrene | |
Sterile | |
21 mm |