Learn More
Sampling Systems™ SteriWare™ Disposable PharmaScoop™
BIOPHARMA
Manufactured in a class 100,000 cleanroom to ensure cleanliness. Sampling Systems™ Polystyrene Disposable Pharma Scoops are individually wrapped and do not require expensive cleaning or cleaning validation.
Brand:  Sampling Systems™ 8045A-250S
 View more versions of this product
Product Code. 16466840
  • 2115.00 NOK / Pack of 85
Only null left
Estimated Shipment: 14-08-2025
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
Disposable Scoop
Individually Bagged
Single-use
White
Polystyrene
250 mL
Sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number122297 Certificate TypeCertificate of Analysis Date16/05/2025 Catalog Number8045A-250S
Lot Number121527 Certificate TypeCertificate of Compliance Date19/03/2025 Catalog Number8045A-250S
Lot Number120258 Certificate TypeCertificate of Compliance Date27/01/2025 Catalog Number8045A-250S
Lot Number119226 Certificate TypeCertificate of Compliance Date23/10/2024 Catalog Number8045A-250S
Lot Number117273 Certificate TypeCertificate of Compliance Date12/02/2024 Catalog Number8045A-250S
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable PharmaScoop™ > Capacity, Metric: 250mL