Learn More
Sampling Systems™ SteriWare™ Disposable PharmaScoop™
Manufactured in a class 100,000 cleanroom to ensure cleanliness. Sampling Systems™ Polystyrene Disposable Pharma Scoops are individually wrapped and do not require expensive cleaning or cleaning validation.
Marke: Sampling Systems™ 8045A-125S
Beschreibung
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Spezifikation
Disposable Scoop | |
Individually Bagged | |
Single-use | |
White |
Polystyrene | |
125 mL | |
Sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
122290 | Certificate of Compliance | 10/06/2025 | 8045A-125S |
121281 | Certificate of Compliance | 09/04/2025 | 8045A-125S |
120249 | Certificate of Compliance | 05/03/2025 | 8045A-125S |
120247 | Certificate of Compliance | 11/11/2024 | 8045A-125S |
120050 | Certificate of Compliance | 23/10/2024 | 8045A-125S |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.