Learn More
Ratiolab™ Polypropylene Urine Beakers
Collect urine specimens in these beakers. Ratiolab™ Polypropylene Urine Beakers are ideal for sampling, transport and storage of solid, pasty and liquid samples.
841.00 NOK - 949.00 NOK
Specifications
Sterility | Non-sterile |
---|---|
Product Type | Urine Beaker |
Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10369182
|
Ratiolab™
5921182 |
125 mL |
841.00 NOK
Pack of 500 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15187073
|
Ratiolab™
5921175 |
60 mL |
949.00 NOK
Pack of 600 |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
Description
- Available in polypropylene (PP) or polystyrene (PS)
- Practical multi-purpose beakers and bottles
- Ideal for the clinical routine
- With red screw-cap
- Supplied in non-sterile or sterile, individually packed
Specifications
Non-sterile | |
Urine Beaker | |
Polypropylene |
Product Title
Ratiolab™ Polypropylene Urine Beakers