Learn More
Cytiva Support Screens for XK and HiScale Columns
For use with XK and HiScale Columns
Brand:  Cytiva 18-9377-01
665.00 NOK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
 View more versions of this product
Product Code. 11560834
  • 700.00 NOK / Pack of 5
In Stock
Explore more special offers
Description
  • Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26 mm
Polypropylene
Documents
One moment while we fetch your results.
Product Title
Cytiva Support Screens for XK and HiScale Columns > I.D.: 26mm