Learn More
Kartell™ Polypropylene Slide Mailer
Ideal for storing and shipping slides. Kartell™ Polypropylene Slide Mailer features a cap with deep threads, which allows a leakproof seal for shipment.
Brand:  Kartell™ 0092200
Product Code. 10584661
  • 499.00 NOK / Pack of 10
Only null left
Estimated Shipment: 14-07-2025
Description
  • Ideal for storing and shipping 5 to 10 microscope slides
  • Unbreakable
  • Chemically inert
  • The container will hold up to 12mL as a speciment vial
  • Easy removal
  • Leakproof seal for shipment
  • Supplied with small loop on cap for tagging
  • Cap should be taped for shipment
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Polypropylene Slide Mailer > HeightMetric: 90mm