Learn More
Fisherbrand™ Polypropylene Racks for Autosampler Vials
Polypropylene vial racks are resistant to most solvents.
257.00 NOK - 271.63 NOK
Specifications
Product Type | Vial Rack |
---|---|
Closure Included | No |
Autoclavable | Autoclavable |
Color | Blue |
Material | Polypropylene |
Product Code | Brand | Dimensions (L x W x H) | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | For Use With (Equipment) | Price | Quantity & Availability | |||||
11767746
|
Fisherbrand™
12 21 2187 |
200 x 105 x 17mm | 1.5mL vials |
271.63 NOK
Each |
In Stock
Log in to see stock. ![]() |
|||||
12672495
|
Fisherbrand™
15 21 2480 |
230 x 117 x 28mm | 4mL Vials |
257.00 NOK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
- Vial-Rack
- 50 positions
- Blue
- Stackable
- Feature alphanumeric indexing for easier vial identification
- Stackable for efficient storage
Specifications
Vial Rack | |
Autoclavable | |
Polypropylene |
No | |
Blue | |
50 |
Product Title
Fisherbrand™ Polypropylene Racks for Autosampler Vials