Learn More
JG Finneran Associates™ Polypropylene Plug Packs
160.00 NOK - 290.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
12357069
|
JG Finneran Associates™
9800-840 |
1 mL |
160.00 NOK
Pack of 100 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
12307079
|
JG Finneran Associates™
9800P-1232 |
2 mL |
290.00 NOK
Pack of 100 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
Specifications
100 |
Product Title
JG Finneran Associates™ Polypropylene Plug Packs