Learn More
Brand™ Polypropylene Microcentrifuge Tube
Brand™ Microcentrifuge Tubes with Lid fit most common rotors and may be spun up to 30,000 RCF.
Brand:  Brand™ 780507
632.90 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10492281
  • 651.00 NOK / Pack of 1000
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Description
  • Free from flash, air bubbles and streaks
  • Uniform wall thickness for consistent, repeatable results
  • Caps for a tight, leak-free seal that is easy to open, even when wearing gloves
  • Simplify sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Locking Lid
Clear
10,000 x g
0.5 mL
1000/Pk.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5126877 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number780507
Product Title
Brand™ Polypropylene Microcentrifuge Tube > Capacity: 0.5mL