Learn More
Masterflex™ Polypropylene Male Luer Fitting
205.00 NOK - 320.32 NOK
Specifications
Material | Polypropylene |
---|---|
For Use With (Application) | Ideal for a variety of applications, including syringes or small tubing |
Packaging Quantity | 25 Pack |
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
15252560
|
Masterflex™
45518-58 |
Fits Male Luer Plug |
243.00 NOK
Pack of 25 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15330491
|
Masterflex™
45508-80 |
Fits Male luer with lock ring x female luer coupler |
320.32 NOK
Pack of 25 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
15353312
|
Masterflex™
45518-44 |
Fits Male Luer x 1/18 in. hose barb |
218.00 NOK
Pack of 25 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
15313322
|
Masterflex™
45518-74 |
Fits Snap Luer Lock Rings |
205.00 NOK
Pack of 25 |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
Specifications
Polypropylene | |
Ideal for a variety of applications, including syringes or small tubing | |
25 Pack |
Product Title
Masterflex™ Polypropylene Male Luer Fitting