Learn More
Brand™ Polypropylene Magnetic Stirring Bar Retrievers
Unique PTFE-coated magnetic stirring bar retrievers that are made with Polypropylene and are ideal for use in most research, scientific, and other types of laboratories.
Brand:  Brand™ 137750
218.78 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 10791561
  • 231.00 NOK / Each
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 27-06-2025
Description
  • Magnetic stirring bar retriever 300 x 10 mm, PP, with ring
  • PTFE-coated 
  • Magnetic core fully sealed
  • L Ă— D 300 mm Ă— 10 mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mm
300 mm
Polypropylene
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brand™ Polypropylene Magnetic Stirring Bar Retrievers > Length: 300mm