Learn More
Brand™ Polypropylene Wide Neck Erlenmeyer Flasks
Flasks with high clarity for visibility of contents during routine laboratory procedures.
472.00 NOK - 646.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10799581
|
Brand™
93172 |
100 mL |
472.00 NOK
Pack of 6 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11888381
|
Brand™
93175 |
250 mL |
641.00 NOK
Pack of 6 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
10465221
|
Brand™
93185 |
1000 mL |
646.00 NOK
Pack of 4 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- Provided with screw cap closure
- With blue graduation marks
- Neck with standard ground socket
- Provided in packs of 6, except for 1000mL flask, which comes in a pack of 4
Specifications
Polypropylene | |
Screw Cap |