Learn More
Buerkle™ Polypropylene Reducing Hose Connectors
Product Code. p-8003072 Shop All Bürkle Products
Click to view available options
For Use With:
4 to 8/12 to 16 mm Hose
4 to 8/8 to 12 mm Hose
8 to 12/12 to 16 mm Hose
Unit Size:
Pack of 10
Product Code. 10642482
Brand: Buerkle™ 87030408
Only null left
Add to basket
Add to basket
Use Buerkle™ Polypropylene Reducing Hose Connectors to make a connections between larger and smaller hoses.
  • Polypropylene
  • Sterilizable
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
For Use With 4 to 8/8 to 12 mm Hose
Packaging Quantity 10/Pk.; 10/Scale
Color White
Product Type Reducing Hose Connector