Learn More
Masterflex™ Polypropylene Barbed Fitting
Adapts from a NPT male thread to a hose barb. Cole-Parmer™ Polypropylene Barbed Fittings are available in a variety of configurations to meet your needs.
Brand:  Masterflex™ 40610-39
 View more versions of this product
Product Code. 12636776
  • 315.00 NOK / Pack of 10
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quick and easy assembly
  • Available in packs of ten
Specifications
Specifications
Barbed Fitting
1/4 NPT Thread to 3/8 in. tubing I.D.
1/4 in. orifice; 1-13/16 x 21/32 in.
ETO or gamma sterilization
Polypropylene
10 Pack
1/4 in. NPT