Learn More
Masterflex™ Polypropylene Barbed Fitting
Adapts from a NPT male thread to a hose barb. Cole-Parmer™ Polypropylene Barbed Fittings are available in a variety of configurations to meet your needs.
Brand: Masterflex™ 40610-42
This item is not returnable.
View return policy
Description
- Quick and easy assembly
- Available in packs of ten
Specifications
Polypropylene | |
10 Pack | |
3/8 in. NPT | |
Barbed Fitting |
3/8 NPT Thread to 3/8 in. tubing I.D. | |
1/4 in. orifice; 1-13/16 x 13/16 in. | |
ETO or gamma sterilization |