Learn More
Masterflex™ Polypropylene Barbed Fitting
Adapts from a NPT male thread to a hose barb. Cole-Parmer™ Polypropylene Barbed Fittings are available in a variety of configurations to meet your needs.
Brand: Masterflex™ 40610-61
This item is not returnable.
View return policy
Description
- Quick and easy assembly
- Available in packs of ten
Specifications
Polypropylene | |
10 Pack | |
3/8 in. NPT | |
Natural |
3/8 NPT Thread x 1/2 in. tubing I.D. | |
11/32 in. orifice; 1-23/32 x 1-23/32 in. | |
NPT male pipe adapter; ETO or gamma sterilization | |
Barbed Fitting |