Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand: Poulten & Graf GmbH 101 09060
This item is not returnable.
View return policy
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of Polypropylene
- Adapter with threads
- Can be used for most chemicals and liquids
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers | |
White | |
Polypropylene | |
1 |
Autoclavable | |
Adapters for OPTIFIX and POLYFIX dispensers | |
OPTIFIX and POLYFIX dispensers | |
GL 60 |