Learn More
Thermo Scientific™ Polymer Tubing Cutter
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using Thermo Scientific™ Polymer Tubing Cutter.
Brand: Thermo Scientific™ A-327
554.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
580.55 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using the Thermo Scientific™ Polymer Tubing Cutter. It is compatible with rigid polymeric tubing and comes with guide holes for 1/16 in. and 1/8 in. tubing.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Tubing Cutter | |
90° Burr-Free End | |
1 Each | |
Tubing |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%