Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Polygram™ polyester sheets
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on Polygram™ polyester sheets come in a variety of sizes. Select fluorescent indicator and choose from a variety of pack sizes.
480.00 NOK - 3170.00 NOK
Specifications
Pore Size | 60 Å |
---|---|
Type | Polyester Sheet |
Fluorescent Indicator | UV254 |
Pore Volume | 0.75 mL/g |
Thickness (Metric) | 0.2 mm |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
10441193
|
Macherey-Nagel™
805023 |
20 x 20 cm |
1765.00 NOK
Pack of 25 |
In Stock
Log in to see stock. |
||||||
10364501
|
Macherey-Nagel™
805021 |
4 x 8 cm |
480.00 NOK
Pack of 50 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10515751
|
Macherey-Nagel™
805017 |
500 x 20 cm |
1685.00 NOK
1 roll |
In Stock
Log in to see stock. |
||||||
10640493
|
Macherey-Nagel™
805022 |
5 x 20 cm |
964.00 NOK
Pack of 50 |
In Stock
Log in to see stock. |
||||||
12864491
|
Macherey-Nagel™
805901 |
2.5 x 7.5 cm |
1780.00 NOK
Pack of 200 |
In Stock
Log in to see stock. |
||||||
10516721
|
Macherey-Nagel™
805024 |
40 x 20 cm |
3170.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
Description
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on Polygram™ polyester sheets come in a variety of sizes. Select a fluorescent indicator and choose from a variety of pack sizes.
Polygram™ polyester sheets for TLC- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ∼ 500 m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 5–17 μm
- Standard grade
- For analytical plates 0.25 mm
- For preparative plates 0.5 and 1 mm
- For 2 mm preparative layers a slightly coarser material is used.
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
60 Å | |
UV254 | |
0.2 mm |
Polyester Sheet | |
0.75 mL/g |